missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572964
Description
Immunogen has 85% identity to chicken Chd7. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.
Chromodomain helicase DNA binding protein 7 (CHD7) is a gene located on chromosome 8q12.2 that encodes a member of the chromodomain helicase DNA-binding (CHD) family, which plays a pivotal role in chromatin remodeling and gene expression regulation. CHD7 contains multiple domains that facilitate its involvement in modifying chromatin structure, thereby influencing the accessibility of DNA for transcription factors and other regulatory proteins. The protein is integral to various developmental processes, including neural crest formation, thus affecting tissues such as the nervous system, skeletal structures, and sensory organs. Mutations in CHD7 are associated with CHARGE syndrome, a multisystem disorder characterized by congenital anomalies including coloboma, heart defects, and ear abnormalities. CHD7's role in chromatin dynamics makes it crucial for normal embryonic development and cellular differentiation. Ongoing research is focused on uncovering CHD7's broader implications in developmental biology and its potential as a therapeutic target in chromatin-related disorders.Identifiants chimiques
| CHD7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| CHD7 | |
| A730019I05Rik; ATP-dependent helicase CHD7; CHARGE association; CHD7; CHD-7; chromodomain helicase DNA binding protein 7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7; CRG; Cycn; Cyn; Dz; Edy; Flo; GENA 47; Gena 52; GENA 60; HH5; IS3; KAL5; KIAA1416; Lda; metis; Mt; Obt; Todo; WBE1; Whi | |
| CHD7 | |
| Primary | |
| Antigen affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% sodium azide | |
| A2AJK6, Q9P2D1 | |
| 320790, 55636 | |
| A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). | |
| Antibody |
Spécifications
| CHD7 | |
| Polyclonal | |
| Unconjugated | |
| CHD7 | |
| A730019I05Rik; ATP-dependent helicase CHD7; CHARGE association; CHD7; CHD-7; chromodomain helicase DNA binding protein 7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7; CRG; Cycn; Cyn; Dz; Edy; Flo; GENA 47; Gena 52; GENA 60; HH5; IS3; KAL5; KIAA1416; Lda; metis; Mt; Obt; Todo; WBE1; Whi | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 320790, 55636 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| A2AJK6, Q9P2D1 | |
| CHD7 | |
| A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |