missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Activated, Norit(R) CA3, Powder, Spectrum™ Chemical

Catalog No. 18609659
Change view
Click to view available options
Quantity:
500 g
2.5 kg
20 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609659 500 g Amber Glass Bottle
18609660 2.5 kg Poly Pail
18609661 20 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609659 Supplier Spectrum Chemical Mfg Cor Supplier No. C1191500GM

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.01
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
IUPAC Name carbon
SMILES [C]

Specifications

CAS Max % 1
pH 2.0 to 3.5
Packaging Amber Glass Bottle
Quantity 500 g
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.