missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Activated, Norit(R) CA1, Powder, Spectrum™ Chemical

Catalog No. 18609662
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609662 500 g Amber Glass Bottle
18609663 2.5 kg Poly Pail
18609664 12 kg Fiber Drum
18609665 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609662 Supplier Spectrum Chemical Mfg Cor Supplier No. C1195500GM

Chemical Identifiers

CAS 7440-44-0

Specifications

CAS Max % 1
pH Approx. 2
Packaging Amber Glass Bottle
Quantity 500 g
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.