missing translation for 'onlineSavingsMsg'
Learn More

CHAPS, White crystalline powder, Hoefer™

Catalog No. 03500504
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03500504 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03500504 Supplier Hoefer Inc Supplier No. GR12110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Critical Micelle Concentration (CMC): 4 to 8 mM

Use in sample preparation for PAGE, IEF, or 2D electrophoresis

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl)-Dimethylammonio]-1-Propane Sulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range ≥98%
Formula Weight 614.88
Assay N analysis
Chemical Name or Material CHAPS
Recommended Storage 4°C, protected from light