missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ CHAPS, Molecular biology grade, Calbiochem™,
$111.00 - $4,580.00
Chemical Identifiers
| CAS | 75621-03-3 |
|---|---|
| Molecular Formula | C32H58N2O7S |
| Molecular Weight (g/mol) | 614.883 |
| InChI Key | UMCMPZBLKLEWAF-GBSSQDIOSA-N |
| Synonym | 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate |
| PubChem CID | 134129639 |
| IUPAC Name | 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate |
| SMILES | CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
2202011GM
|
MilliporeSigma™
2202011GM |
1 g | Glass Bottle |
Each for $111.00
|
|
||||
|
2202015GM
|
MilliporeSigma™
2202015GM |
5 g | Glass Bottle |
Each for $322.00
|
|
||||
|
22020110GM
|
MilliporeSigma™
22020110GM |
10 g | Glass Bottle |
Each for $547.00
|
|
||||
|
22020125GM
|
MilliporeSigma™
22020125GM |
25 g | Glass Bottle |
Each for $1,060.00
|
|
||||
|
220201250GM
|
MilliporeSigma™
220201250GM |
250 g | Glass Bottle |
Each for $3,380.00
|
|
||||
|
2202011KG
|
MilliporeSigma™
2202011KG |
1 kg | Glass Bottle |
Each for $4,580.00
|
|
||||
Chemical Identifiers
| 75621-03-3 | |
| 614.883 | |
| 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate | |
| 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate |
| C32H58N2O7S | |
| UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
| 134129639 | |
| CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C |
Specifications
| White | |
| C32H58N2O7S | |
| UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
| 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate | |
| 134129639 | |
| Molecular Biology |
| 75621-03-3 | |
| 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate | |
| CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C | |
| 614.883 | |
| ≥98% (NMR) |