missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ CHAPS, Molecular biology grade, Calbiochem™,
Supplier: MilliporeSigma™ 22020110GM
| Quantity | 10 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 75621-03-3 | |
| 614.883 | |
| 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate | |
| 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate |
| C32H58N2O7S | |
| UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
| 134129639 | |
| CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C |
Specifications
| White | |
| 10 g | |
| C32H58N2O7S | |
| UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
| 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate | |
| 134129639 | |
| Molecular Biology |
| 75621-03-3 | |
| Glass Bottle | |
| 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate | |
| CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C | |
| 614.883 | |
| ≥98% (NMR) |