missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CHAPS, Molecular biology grade, Calbiochem™,

Catalog No. 220201250GM
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
250 g
1 kg
Packaging:
Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
220201250GM 250 g Glass Bottle
2202011GM 1 g Glass Bottle
2202015GM 5 g Glass Bottle
22020110GM 10 g Glass Bottle
22020125GM 25 g Glass Bottle
2202011KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 220201250GM Supplier MilliporeSigma™ Supplier No. 220201250GM

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Color White
Quantity 250 g
Packaging Glass Bottle
Percent Purity ≥98% (NMR)
Grade Molecular Biology