missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CHAPS, Molecular biology grade, Calbiochem™,

Supplier:  MilliporeSigma™ 22020125GM

Quantity 25 g
Packaging Glass Bottle

 View more versions of this product

Catalog No. 22020125GM


Only null left

Description

Description

Specifications

Chemical Identifiers

75621-03-3
614.883
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
C32H58N2O7S
UMCMPZBLKLEWAF-GBSSQDIOSA-N
134129639
CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

White
25 g
C32H58N2O7S
UMCMPZBLKLEWAF-GBSSQDIOSA-N
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
134129639
Molecular Biology
75621-03-3
Glass Bottle
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
614.883
≥98% (NMR)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions