Learn More
Stemcell Technologies mTeSR™1 DFS Item
Supplier:  Stemcell Technologies Inc 85850
Use this specialized, feeder-free culture medium to achieve more consistent human pluripotent stem cell cultures with homogenous, undifferentiated phenotypes. Manufactured under relevant cGMPs, mTeSR™1 ensures the highest quality and consistency for reproducible results in your fundamental research, as well as for cell therapy and investigational new drug research applications. This serum-free, complete cell culture medium is made with pre-screened raw materials to ensure batch-to-batch consistency and robust performance in feeder-free hPSC culture. Use established protocols for applications ranging from derivation to differentiation with this most widely published feeder-free hPSC culture medium, which has been used by leading pluripotent stem cell researchers to successfully maintain thousands of hPSC lines in over 50 countries.
Catalog No. NC1274780
Only null left
Explore available promotions