Learn More
Sterlitech Corporation CF042 Cell Assembly, Crossflow, 316 SS (CF042SS)
Numéro de catalogue. 501952345
Encompass
The Sterlitech CF042 membrane cell is a lab scale cross flow filtration unit designed to provide fast and accurate performance data with minimal amounts of product, expense, and time.
CF042 Membrane Cell Product Availability:
  • Cross flow cell units available in Delrin, PTFE, Stainless Steel, or Acrylic 
  • 1000 psi (69 bar) max operating pressure (PTFE and Acrylic versions are 400 psi)
  • Active membrane area: 42cm2
Please note: A feed flow pump is required to operate the CF042 cross flow cell unit. We recommend the Hydracell pump by Wanner Engineering, which is a positive displacement, diaphragm pump designed for high pressure, lower flow applications. When ordering, be sure to specify the power requirements for the motor. The minimum power available is 110V, 60 Hz. 220 V and 50 Hz motors are also available. There may be an additional cost if a variable speed drive is required. 
Numéro de catalogue. 501952345 Fournisseur Sterlitech Corporation Code fournisseur 1160236
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Sterlitech CF042 membrane cell is a lab scale cross flow filtration unit designed to provide fast and accurate performance data with minimal amounts of product, expense, and time.
CF042 Membrane Cell Product Availability:
  • Cross flow cell units available in Delrin, PTFE, Stainless Steel, or Acrylic 
  • 1000 psi (69 bar) max operating pressure (PTFE and Acrylic versions are 400 psi)
  • Active membrane area: 42cm2
Please note: A feed flow pump is required to operate the CF042 cross flow cell unit. We recommend the Hydracell pump by Wanner Engineering, which is a positive displacement, diaphragm pump designed for high pressure, lower flow applications. When ordering, be sure to specify the power requirements for the motor. The minimum power available is 110V, 60 Hz. 220 V and 50 Hz motors are also available. There may be an additional cost if a variable speed drive is required.