missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethylammonium bromide, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219502990

Quantity 500 g

 Afficher plus de versions de ce produit

Numéro de catalogue. ICN19502990


Il en reste null

Description

Description

Spécifications

Identifiants chimiques

57-09-0
364.46
LZZYPRNAOMGNLH-UHFFFAOYSA-M
5974
hexadecyltrimethylazanium bromide
C19H42BrN
MFCD00011772
Hexadecyltrimethylammonium bromide, CTAB
CHEBI:3567
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Spécifications

Cetyltrimethylammonium bromide
57-09-0
500 g
C19H42BrN
Hexadecyltrimethylammonium bromide, CTAB
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
364.46
CHEBI:3567
White
Powder
0.9906
MFCD00011772
LZZYPRNAOMGNLH-UHFFFAOYSA-M
hexadecyltrimethylazanium bromide
5974
364.5
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

EINECSNumber : 200-311-3

Recommended Storage : Store at Room Temperature (15 to 30°C).

FDS
Documents

Documents

Promotions

Promotions