missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethylammonium bromide, ∽98%, Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19400480
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19400480 100 g
ICN19400483 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19400480 Supplier MP Biomedicals Inc Supplier No. 0219400480

Chemical Identifiers

CAS 57-09-0
Molecular Formula C19H42BrN
Molecular Weight (g/mol) 364.46
MDL Number MFCD00011772
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
Synonym cetrimonium bromide, cetyltrimethylammonium bromide, hexadecyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
PubChem CID 5974
ChEBI CHEBI:3567
IUPAC Name hexadecyl(trimethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color White
Quantity 100 g
Formula Weight 364.5
Percent Purity ∼98%
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Cetyltrimethylammonium bromide