missing translation for 'onlineSavingsMsg'
Learn More

Cetylpyridinum chloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019638
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019638 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019638 Supplier Merck Emd Millipore Supplier No. PHR12261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cetylpyridinum chloride is a quaternary ammonium compound and an active ingredient of antiseptic oral mouth rinses with broad spectrum antimicrobial activity. Its mode of action involves a rapid bactericidal effect on Gram-positive pathogens and a fungicide effect on certain yeasts.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 6004-24-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C21H38 ClN · H2O
MDL Number MFCD00149977
Melting Point 83°C to 86°C
Synonym Cetylpyridinium chloride monohydrate; 1-Hexadecylpyridinium chloride; Hexadecylpyridinium chloride monohydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 358.00
Grade Certified Reference Material
Show More Show Less