missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetostearyl Alcohol, NF, 90-102%, Spectrum™ Chemical
C1888, 8005-44-5
$563.85 - $10157.33
Chemical Identifiers
| CAS | 8005-44-5 |
|---|---|
| Molecular Formula | C34H72O2 |
| Molecular Weight (g/mol) | 512.95 |
| InChI Key | UBHWBODXJBSFLH-UHFFFAOYSA-N |
| IUPAC Name | hexadecan-1-ol; octadecan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601549
|
Spectrum Chemical Mfg Cor
C1888500GM |
500 g | Poly Bottle |
Each for $563.85
|
|
||||
|
18601550
|
Spectrum Chemical Mfg Cor
C188825KG |
2.5 kg | Poly Pail |
Each for $1,586.30
|
|
||||
|
18601552
|
Spectrum Chemical Mfg Cor
C188845KGBL |
45 kg | Fiber Drum |
Each for $10,157.33
|
|
||||
Description
Spectrum™ Chemical Cetostearyl Alcohol, NF is used as a stabilizer and opacifying agent. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 8005-44-5 | |
| 512.95 | |
| hexadecan-1-ol; octadecan-1-ol |
| C34H72O2 | |
| UBHWBODXJBSFLH-UHFFFAOYSA-N | |
| CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO |
Specifications
| 8005-44-5 | |
| Poly Bottle | |
| 500 g | |
| CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO | |
| 512.95 | |
| NF |
| 100% | |
| C34H72O2 | |
| UBHWBODXJBSFLH-UHFFFAOYSA-N | |
| hexadecan-1-ol; octadecan-1-ol | |
| 90-102% |