missing translation for 'onlineSavingsMsg'
Learn More

Cetostearyl Alcohol, NF, 90-102%, Spectrum™ Chemical

Catalog No. 18601552
Change view
Click to view available options
Quantity:
500 g
2.5 kg
45 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601552 45 kg Poly Bottle
18601549 500 g Poly Bottle
18601550 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601552 Supplier Spectrum Chemical Mfg Cor Supplier No. C188845KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1888, 8005-44-5

Spectrum™ Chemical Cetostearyl Alcohol, NF is used as a stabilizer and opacifying agent. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8005-44-5
Molecular Formula C34H72O2
Molecular Weight (g/mol) 512.95
InChI Key UBHWBODXJBSFLH-UHFFFAOYSA-N
IUPAC Name hexadecan-1-ol; octadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 45 kg
Percent Purity 90-102%
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Cetostearyl Alcohol