accessibility menu, dialog, popup

UserName

H3COHH3COH

CAS RN 8005-44-5

IUPAC Name: hexadecan-1-ol; octadecan-1-ol
Synonyms: cetostearyl alcohol
Molecular Weight (g/mol): 512.95
Molecular Formula: C34H72O2
InChi Key: UBHWBODXJBSFLH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)