missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetostearyl Alcohol, NF, 90-102%, Spectrum™ Chemical
Description
Spectrum™ Chemical Cetostearyl Alcohol, NF is used as a stabilizer and opacifying agent. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Percent Purity | 90-102% |
| Grade | NF |