missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine Dihydrochloride 98.0+%, TCI America™

Catalog No. C23165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23165G 5 g
C231625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23165G Supplier TCI America Supplier No. C23165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83881-52-1
Molecular Formula C21H27Cl3N2O3
Molecular Weight (g/mol) 461.81
MDL Number MFCD00941428
InChI Key PGLIUCLTXOYQMV-UHFFFAOYNA-N
Synonym cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler
PubChem CID 55182
IUPAC Name dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride
SMILES [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 225°C
Color White
Quantity 5 g
Formula Weight 461.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cetirizine Dihydrochloride
RTECSNumber AG0980000
TSCA No