missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine dihydrochloride, 99+%

Catalog No. AAJ6354906
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6354906 5 g
AAJ6354909 10 g
AAJ6354914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6354906 Supplier Thermo Scientific Chemicals Supplier No. J6354906

CAS: 83881-52-1 | C21H27Cl3N2O3 | 461.81 g/mol

Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.

Solubility
Soluble in water, DMSO, ethanol, and methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 83881-52-1
Molecular Formula C21H27Cl3N2O3
Molecular Weight (g/mol) 461.81
MDL Number MFCD00941428
InChI Key PGLIUCLTXOYQMV-UHFFFAOYNA-N
Synonym cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler
PubChem CID 55182
IUPAC Name dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride
SMILES [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1

Specifications

Color White
Quantity 5 g
Merck Index 14,2022
Solubility Information Soluble in water,DMSO,ethanol,and methanol.
Formula Weight 461.82
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Cetirizine dihydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber AG0980000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only