missing translation for 'onlineSavingsMsg'
Learn More

Cesium trifluoroacetate

Catalog No. AA4463318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4463318 50 g
AA4463309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4463318 Supplier Thermo Scientific Chemicals Supplier No. 04463318
Only null left

CAS: 21907-50-6 | C2CsF3O2 | 245.92 g/mol

It is used in acid catalyzed reactions, especially those where an ester is cleaved in peptide synthesis. Used as an ion pairing agent in liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21907-50-6
Molecular Formula C2CsF3O2
Molecular Weight (g/mol) 245.92
MDL Number MFCD00051304
InChI Key RJYSYRSELCQCSO-UHFFFAOYSA-M
Synonym cesium trifluoroacetate, caesio 2,2,2-trifluoroacetate, trifluoroacetic acid cesium salt, cesium trifluoroacetate solution, cstfa, caesium trifluoroacetate, cesium 2,2,2-trifluoroacetate, caesium 1+ trifluoroacetate, cesium 2,2,2-tris fluoranyl ethanoate, caesium 1+ ion trifluoroacetate
PubChem CID 23689558
IUPAC Name cesium;2,2,2-trifluoroacetate
SMILES [Cs+].[O-]C(=O)C(F)(F)F

Specifications

Melting Point 115°C
Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in water (2 M,clear,colorless) at 20°C. Soluble in organic solvents.
Formula Weight 245.92
Chemical Name or Material Cesium trifluoroacetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only