missing translation for 'onlineSavingsMsg'
Learn More

Cesium sulfate, 99%

Catalog No. AAA1676714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1676714 25 g
AAA1676722 100 g
AAA1676736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1676714 Supplier Thermo Scientific Chemicals Supplier No. A1676714
Only null left

CAS: 10294-54-9 | Cs2O4S | 361.87 g/mol

Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium sulfate is used to prepare dense aqueous solutions for use in isopycnic centrifugation. It is used as metal oxide catalyst enhancer for heterogeneous processes.

Solubility
Soluble in water. (1820 g/L) at 20°C

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water, reducing agents and oxidizing agents.

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate
PubChem CID 25137
IUPAC Name dicesium;sulfate
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Melting Point 1,010°C
Quantity 25 g
Merck Index 14,2016
Solubility Information Soluble in water. (1820g/L) at 20°C
Formula Weight 361.87
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.24
Chemical Name or Material Cesium sulfate
Hazard Category H302-H361f-H373
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 233-662-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only