missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Sulfate, 99.5%, Spectrum™ Chemical
Description
- Caesium salt of sulfuric acid used to prepare dense aqueous solutions for isopycnic centrifugation.
Specifications
Specifications
| Melting Point | 1010°C |
| Color | Colorless |
| CAS Max % | 1 |
| Quantity | 5 g |
| Assay Percent Range | 99.5% |
| Synonym | Caesium Sulfate |
| Solubility Information | Easily Soluble in Cold Water, Insoluble in Methanol, Acetone |
| Formula Weight | 361.87 g/mol |
| Percent Purity | 99.5% |
| Grade | Reagent |
| Show More |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.