missing translation for 'onlineSavingsMsg'
Learn More

Cesium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLCS23X

 View more versions of this product

Catalog No. PLCS2-3X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cs10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Cesium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP
Certified Reference Material
[N+](=O)([O-])[O-].[Cs+]
cesium;nitrate
500 mL
Assurance
Triple-leached LDPE bottle
7789-18-6,7697-37-2,7732-18-5
CsNO3
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
NLSCHDZTHVNDCP-UHFFFAOYSA-N
5% HNO3
194.909
62674

Chemical Identifiers

7789-18-6
194.909
62674
[N+](=O)([O-])[O-].[Cs+]
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only