missing translation for 'onlineSavingsMsg'
Learn More

Cesium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$488.06 - $1087.81

Chemical Identifiers

CAS 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLCS2-3Y
View Documents
SPEX CertiPrep Group LLC
PLCS23Y
125 mL Triple-leached LDPE bottle
Each for $488.06
Only null left
 
PLCS2-3X
SDS View Documents
SPEX CertiPrep Group LLC
PLCS23X
500 mL Triple-leached LDPE bottle
Each for $1,087.81
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cs10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7789-18-6
194.909
62674
[N+](=O)([O-])[O-].[Cs+]
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate

Specifications

Triple-leached LDPE bottle
7789-18-6,7697-37-2,7732-18-5
CsNO3
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
NLSCHDZTHVNDCP-UHFFFAOYSA-N
5% HNO3
194.909
62674
Cesium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP
Certified Reference Material
[N+](=O)([O-])[O-].[Cs+]
cesium;nitrate
125 mL
Assurance
Videos
Safety and Handling

Safety and Handling

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only