missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Pivalate 97.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 348°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 234.03 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cesium Pivalate |
Sécurité et manipulation
| TSCA | No |