missing translation for 'onlineSavingsMsg'
Learn More

Cesium Iodide 99.0+%, TCI America™

Catalog No. C220525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C220525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C220525G Supplier TCI America Supplier No. C220525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical reagent suitable for all laboratory applications

  • Cesium Iodide is a high-purity laboratory reagent

Chemical Identifiers

CAS 7789-17-5
Molecular Formula CsI
Molecular Weight (g/mol) 259.81
MDL Number MFCD00010962
InChI Key XQPRBTXUXXVTKB-UHFFFAOYSA-M
Synonym cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
PubChem CID 24601
IUPAC Name caesium(1+) iodide
SMILES [I-].[Cs+]

Specifications

Melting Point 632°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range ≥99.0%
Merck Index 2014
Solubility Information Soluble in Water, Soluble in Ethanol, Methanol, Insoluble in Acetone
Formula Weight 259.81
Percent Purity ≥99.0% (T)
Manufacturing Origin USA
Sensitivity Hygroscopic
Appearance White Powder to Crystal
Vapor Pressure 0.00045 kPa at 25°C
Decomposition Information Hydrogen Halides, Metal Oxides
Chemical Name or Material Cesium Iodide
Show More Show Less
EINECSNumber (1)-1063
RTECSNumber FL0350000
TSCA Yes