missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Iodide, Approx. 99.9%, Spectrum™ Chemical
Description
- Suitable for general industrial or research use
- Used as an input phosphor in x-ray image intensifier tubes for fluoroscopy equipment
- Supplied by spectrum are indicative of a grade suitable for general industrial use or research purposes
- Not suitable for human consumption or therapeutic use
Specifications
Specifications
| Boiling Point | 1280°C |
| Melting Point | 626°C |
| Color | White |
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 5 kg |
| Assay Percent Range | 99.90% |
| Molecular Formula | CsI |
| Solubility Information | Soluble in cold Water |
| Molecular Weight (g/mol) | 259.81 |
| Show More |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.