missing translation for 'onlineSavingsMsg'
Learn More

Cesium ICP Standard, 10,000 ppm Cs in 5% HNO3, Ricca Chemical

Catalog No. PCS10KN100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PCS10KN100 100 mL Natural LDPE Bottle
PCS10KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PCS10KN100 Supplier Ricca Chemical Company Supplier No. PCS10KN100

Chemical Identifiers

CAS 7789-18-6 , 7697-37-2 , 7789-18-6
Molecular Formula CsNO3
Molecular Weight (g/mol) 194.909
InChI Key NLSCHDZTHVNDCP-UHFFFAOYSA-N
PubChem CID 62674
IUPAC Name cesium;nitrate
SMILES [N+](=O)([O-])[O-].[Cs+]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Cs
CAS Min % 90.0, 6.76, 1.32
CAS Max % 94.0, 7.03, 1.37
Equivalence 1 mL = 10 mg Cs
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 100 mL
Chemical Name or Material Cesium ICP Standard
Show More Show Less