missing translation for 'onlineSavingsMsg'
Learn More

Cesium Fluoride, Optical Grade, 99%, Spectrum™ Chemical

$326.11 - $33028.28

Chemical Identifiers

CAS 13400-13-0
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609633
View Documents
Spectrum Chemical Mfg Cor
CE1185GM
5 g Amber Glass Bottle
Each for $326.11
Only null left
 
18609634
View Documents
Spectrum Chemical Mfg Cor
CE11825GM
25 g Amber Glass Bottle
Each for $751.32
Only null left
 
18609635
View Documents
Spectrum Chemical Mfg Cor
CE118100GM
100 g Amber Glass Bottle
Each for $1,898.31
Only null left
 
18609636
View Documents
Spectrum Chemical Mfg Cor
CE11825KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13400-13-0

Specifications

13400-13-0
5 g
Optical
1
99%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.