missing translation for 'onlineSavingsMsg'
Learn More

Cesium Fluoride, Optical Grade, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor CE11825GM

 View more versions of this product

Catalog No. 18609634


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

13400-13-0
25 g
Optical
1
99%
Amber Glass Bottle

Chemical Identifiers

13400-13-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.