missing translation for 'onlineSavingsMsg'
Learn More

Cesium Fluoride, Optical Grade, 99%, Spectrum™ Chemical

Catalog No. 18609633
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
25 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609633 5 g Amber Glass Bottle
18609634 25 g Amber Glass Bottle
18609635 100 g Amber Glass Bottle
18609636 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609633 Supplier Spectrum Chemical Mfg Cor Supplier No. CE1185GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optical grade chemical in fluoride anion source and suitable for crystal growing applications

  • Used in organic synthesis and suitable for laboratory applications
  • Suitable for inorganic synthesis of fluoride ions

Chemical Identifiers

CAS 13400-13-0

Specifications

Boiling Point 685°C
Melting Point 29°C
Color Gold, Silver-gold
CAS Max % 1
Physical Form Powder, Crystals and/or Chunks
Quantity 5 g
Assay Percent Range 99%
Molecular Formula CsF
UN Number UN3288
SMILES [Cs+].[F-]
Molecular Weight (g/mol) 151.91
Formula Weight 151.91 g/mol
Percent Purity 99%
Grade Optical
Packaging Amber Glass Bottle
Appearance Powder
Chemical Name or Material Cesium Fluoride
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.