missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Fluoride, Optical Grade, 99%, Spectrum™ Chemical
Description
- Used in organic synthesis and suitable for laboratory applications
- Suitable for inorganic synthesis of fluoride ions
Specifications
Specifications
| Boiling Point | 1251°C |
| CAS Max % | 1 |
| Physical Form | Powder, Crystals and/or Chunks |
| Quantity | 25 kg |
| Assay Percent Range | 99% |
| Molecular Formula | CsF |
| UN Number | UN3288 |
| Synonym | Caesium Fluoride |
| Solubility Information | Soluble in Water |
| Molecular Weight (g/mol) | 151.91 |
| Show More |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.