missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Cesium Chloride, Molecular Biology Grade, Calbiochem™,
Description
Hygroscopic
Specifications
Specifications
| Quantity | 500 g |
| Assay Percent Range | ≥99% |
| Solubility Information | Easily soluble in cold water |
| Formula Weight | 168.4 |
| Grade | by titration (argentometry) |
| Product Line | CalbioChem™ |
| Chemical Name or Material | Cesium Chloride |
Safety and Handling
| EINECSNumber | 231-600-2 |
| RTECSNumber | FK9625000 |
| Recommended Storage | Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F). |