missing translation for 'onlineSavingsMsg'
Learn More

Cesium Acetate, Ultrapure, 99.99%, Spectrum™ Chemical

Catalog No. 18609617
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609617 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18609617 Supplier Spectrum Chemical Mfg Cor Supplier No. C1428100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 3396-11-0

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Percent Purity 0.9999
Grade Ultrapure

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.