missing translation for 'onlineSavingsMsg'
Learn More

Cerium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLCE22X

 View more versions of this product

Catalog No. PLCE2-2X


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ce1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Cerium Standard
7440-45-1 , 7697-37-2 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
1.0,5.0,94.0
MFCD00010924
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
GWXLDORMOJMVQZ-UHFFFAOYSA-N
2% HNO3
140.12
23974
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
1.0,5.0,94.0
Ce
AA, ICP
Certified Reference Material
[Ce]
cerium
500 mL
CHEBI:33369
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLCE2-2Y, 2X    10,000    PLCE2-3Y, -3X

Chemical Identifiers

7440-45-1
140.12
GWXLDORMOJMVQZ-UHFFFAOYSA-N
CHEBI:33369
[Ce]
Ce
MFCD00010924
23974
cerium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only