missing translation for 'onlineSavingsMsg'
Learn More

Cerium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

Catalog No. PLCE23Y
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PLCE23Y 125 mL Triple-leached LDPE bottle
PLCE23X 500 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLCE23Y Supplier SPEX CertiPrep Group LLC Supplier No. PLCE23Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ce10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-45-1 , 7697-37-2 , 7732-18-5
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 5% HNO3
Quantity 125 mL
Grade Assurance
Chemical Name or Material Cerium Standard
Hazard Category Causes skin irritation, serious eye irritation
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only