missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) Oxide, 1 to 3mm, For Elementary Analysis, MilliporeSigma™

Catalog No. M1022630100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1022630100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1022630100 Supplier Technidata Supplier No. 1.02263.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O

Specifications

Physical Form Solid
Quantity 100 g
Formula Weight 172.11. (molar mass)
Odor Odorless
Packaging Plastic can
Appearance Light gray solid
Chemical Name or Material Cerium(IV) oxide