missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium ICP Standard, 10,000 ppm Ce in 10% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 10,000ppm Ce |
| CAS Min % | 83.0, 12.47, 2.57 |
| CAS Max % | 86.0, 12.98, 2.68 |
| Equivalence | 1 mL = 10 mg Ce |
| Chemical Name or Material | Cerium ICP Standard |