missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Meperidine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Numéro de catalogue. 01001585
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01001585 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01001585 Fournisseur Merck Emd Millipore Code fournisseur M0351ML
Il en reste null

Certified reference material

Meperidine is an opioid analgesic commonly known by its brand name, Demerol™. This certified solution standard is applicable for use in urine drug testing, pain prescription monitoring, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS. This painkiller, considered to have a fast onset of action, has a history of recreational abuse.

Spécifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol
Linear Formula C15H21NO2
MDL Number MFCD00057376
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 247.33

CERILLIANT is a registered trademark of Cerilliant Corporation