missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceric Sulfate, 0.100 N (N/10) in 2 N Sulfuric Acid, Ricca Chemical
Specifications
Specifications
| CAS | 13590-82-4, 7664-93-9, 7732-18-5 |
| Molecular Formula | CeO8S2 |
| MDL Number | MFCD00148852 |
| Synonym | ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate |
| InChI Key | VZDYWEUILIUIDF-UHFFFAOYSA-J |
| SMILES | [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| IUPAC Name | λ⁴-cerium(4+) disulfate |
| Molecular Weight (g/mol) | 332.23 |
| PubChem CID | 159684 |
| Packaging | Cubitainer |
| Show More |