missing translation for 'onlineSavingsMsg'
Learn More

Ceric Sulfate, 0.100 N (N/10) in 2 N Sulfuric Acid, Ricca Chemical

Catalog No. 190032
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
190032 1 L Natural Poly Bottle
190016 500 mL Natural Poly Bottle
19001 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 190032 Supplier Ricca Chemical Company Supplier No. 190032
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13590-82-4 , 7664-93-9 , 7732-18-5
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
PubChem CID 159684
IUPAC Name λ4-cerium(4+) disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Orange
Packaging Natural Poly Bottle
Solvent or Matrix 2N Sulfuric Acid
pH <1
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.100N
Grade Laboratory
Chemical Name or Material Ceric Sulfate