missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Sulfate, in 1.0N Sulfuric Acid, Certified, 0.100N ±0.001N (0.10M), LabChem™

Catalog No. LC129306
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass
Cubitainer™
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC129306 10 L Cubitainer™
LC129301 500 mL Amber Glass
LC129302 1 L Amber Glass
LC129304 4 L Amber Glass
LC129305 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC129306 Supplier LabChem Supplier No. LC129306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16774-21-3 | CeH8N8O18 | 548.22 g/mol

Chemical Identifiers

CAS 16774-21-3 , 7732-18-5 , 7664-93-9
Molecular Formula CeH8N8O18
Molecular Weight (g/mol) 548.22
InChI Key WIBGOERAEYJBOT-UHFFFAOYSA-N
Synonym Ceric ammonium nitrate, CAN
PubChem CID 24181046  
IUPAC Name λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
SMILES N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Orange
Concentration 0.100N ±0.001N (0.10M)
CAS Max % 5.7, 91.3, 3
Identification Passes Test
Packaging Cubitainer™
Quantity 10 L
Physical Form Liquid
Linear Formula Ce(NH4)2(NO3)6
UN Number UN2796
Solubility Information Soluble in water
Formula Weight 548.23
Grade Certified
Concentration or Composition (by Analyte or Components) 0.100N ±0.001N (0.10M)
Chemical Name or Material Ceric Ammonium Sulfate, in 1.0N Sulfuric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature