missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cephalothin (acid), 97%

Catalog No. AAH3617514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3617514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3617514 Supplier Thermo Scientific™ Supplier No. H3617514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 153-61-7
Melting Point 160-161°
Molecular Formula C16H15N2NaO6S2
MDL Number MFCD00242614
Quantity 25 g
InChI Key VUFGUVLLDPOSBC-XRZFDKQNSA-M
SMILES [Na+].CC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CC=CS1)C2=O)C([O-])=O
IUPAC Name sodium (6R,7R)-3-[(acetyloxy)methyl]-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol) 418.41
Formula Weight 396.43
Percent Purity 97%
Physical Form Solid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 205-815-7
RTECSNumber XI0388000