missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CEP
Description
CEP is a chemoproteomic reagent. It is a clickable electrophilic purine used to directly quantify protein-RNA interactions on proteins through photoaffinity competition with 4-thiouridine (4SU)-labeled RNA in cells. It is a cysteine reactive probe used in photo-activatable-competition and chemoproteomic enrichment (PACCE) method for global quantification of protein-RNA interactions in living cells.
Spécifications
Spécifications
| Chemical Name or Material | 2,6-Dichloro-7-(2-propyn-1-yl)-7H-purine |
| CAS | 1838570-66-3 |
| Quantity | 5 mg |
| Target | DNA, RNA and Protein Synthesis Compounds |
| Molecular Formula | C8H4Cl2N4 |
| Purity | 0.98 |