missing translation for 'onlineSavingsMsg'
Learn More

Labconco CentriVap™ Pro with Glass Lid and Heat Boost
GREENER_CHOICE

Vacuum concentrator that is designed to rapidly concentrate multiple small samples using centrifugal force, vacuum and heat.

Supplier:  Labconco 8511200

 Afficher plus de versions de ce produit

Numéro de catalogue. 12199503

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

  • Compact, bench top footprint
  • Pressure, temperature, rotor speed and run time can be easily set with the touch of a few buttons and stored in the methods menu. User may also alter all run parameters while the program is in progress.
  • Easily view, plot, and export sample run data for analysis and publication. CSV and PDF file formats supported.
  • Large chamber accommodates a variety of rotors including a selection that can be double stacked for added capacity up to 312 sample tubes or 6 microtiter plates per run.
  • Customize run parameters, record and download sample data, set up automated device alerts through Ethernet, and configure device settings on an integrated 5 in. full color touch screen.
  • Additional 300 W Heat Boost speeds chamber heating for faster sample processing and increased performance.
  • Built in methods menu for processing common laboratory sample types. Includes modes for small-scale lyophilization and vacuum desiccation.
Spécifications

Identifiants chimiques

Epoxy-coated Aluminum
Ambient to 100°C
North American
Vacuum Concentrator
CentriVap Pro
Up to 50 mL
No

Spécifications

Epoxy-coated Aluminum
Ambient to 100°C
North American
Vacuum Concentrator
CentriVap Pro
Up to 50 mL
No
115 V
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions