Learn More
Thermo Scientific™ CentriPAK BPC Sterilized Single-use for Sorvall BIOS 16 Bioprocessing Centrifuge
Thermo Scientific™ CentriPAK™ BioProcess Container (BPC), designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor, is providing a single-use, sterilized, closed system for cell culture separations in the bioprocessing industry.
Specifications
| For Use With (Application) | For mammalian, yeast and bacterial cell separations |
|---|---|
| Sterility | Sterile |
| Product Line | Polaris |
| Shipping Condition | Packages placed in strong cardboard cartons with product and lot identification |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|
75003885
|
Thermo Scientific™
75003885 |
CentriPAK BPC Single |
N/A
|
|
|||||
|
75003890
|
Thermo Scientific™
75003890 |
CentriPAK BPC Steam-through Adapter |
N/A
|
|
|||||
|
75003891
|
Thermo Scientific™
75003891 |
CentriPAK BPC Single with Quick Connect |
N/A
|
|
|||||
|
75003883
|
Thermo Scientific™
75003883 |
CentriPAK Labtainer™ BPC |
N/A
|
|
|||||
|
75003880
|
Thermo Scientific™
75003880 |
CentriPAK BPC |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Thermo Scientific™ CentriPAK™ BioProcess Container (BPC) chambers, manifold, labtainer (sterilized) with connection. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
Thermo Scientific™ CentriPAK™ BioProcess Container (BPC) enables you to:
- Reduce cross-contamination risk while separating cell cultures
- Perform gentle, high-throughput and high-efficiency harvesting through centrifugation
- Eliminate post-use cleaning steps required with reusable containers
- Meet rigorous quality control and assurance practices for the manufacture of industry-standard material components and aseptic product contact film
Specifications
| For mammalian, yeast and bacterial cell separations | |
| Polaris | |
| Single-use | |
| Sorvall BIOS 16 Bioprocessing Centrifuge |
| Sterile | |
| Packages placed in strong cardboard cartons with product and lot identification | |
| Flexible Bioprocess Container | |
| Not Autoclavable |