missing translation for 'onlineSavingsMsg'
Learn More

OHAUS Frontier™ Centrifuge Multi

Catalog No. 01920168
Change view
Click to view available options
Capacity:
4 x 15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
01920168 4 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01920168 Supplier OHAUS Supplier No. OHS83041073

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Frontier™ multi centrifuges are reliable compact low-speed general purpose centrifuges. The standard rotor options can process both round and conical bottom tubes (including common blood tubes), and can also be adapted easily with accessories to process smaller tubes.

  • Designed for ease of operation, Frontier™ Multi Centrifuges feature convenient control on an intuitive interface, which allows for responsive operation, even when wearing gloves.
  • Frontier™ Multi Centrifuges can accommodate standard 50 mL and 15 mL tubes, including common blood tubes.
  • When used with accessories, rotors can be adapted for sizes as small as 1.5 mL.
  • Enclosing all its functionalities within a small footprint, Frontier™ Multi Centrifuges save valuable space on the lab bench.

Specifications

Capacity 4 x 15 mL
Max. RCF 4,445 x g
Max. Speed 6,800 rpm
Product Type Centrifuge