missing translation for 'onlineSavingsMsg'
Learn More

Cellulose DS-0, Powder, For thin layer chromatography, MilliporeSigma™ Supelco™

Catalog No. 111015794
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015794 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015794 Supplier Merck Emd Millipore Supplier No. 22197250GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00081512
Synonym Cellulose powder; Cotton linters

Specifications

Physical Form Powder
CAS 9004-34-6
Quantity 250 g
Grade TLC
RTECSNumber FJ5691460