missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose acetate, average M.W. 100,000

Catalog No. AC177780250
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177780250 25 g Plastic bottle
AC177785000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177780250 Supplier Thermo Scientific Chemicals Supplier No. 177780250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 9004-35-7 | (C6H7O5)nC2H3O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-35-7
Molecular Formula (C6H7O5)nC2H3O
MDL Number MFCD00081496

Specifications

Color White
Physical Form Powder
Merck Index 14, 1966
Quantity 25 g
Formula Weight 100,000 (Average )
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Plastic bottle
Chemical Name or Material Cellulose acetate

RUO – Research Use Only