Learn More
Celltreat Scientific Prod 10mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile
Supplier: Celltreat Scientific Products 229210B
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification.